Learn More
Beckman Coulter POLY TUBES 11X34MM 50/PK
Catalog No. NC9780632
Encompass_Preferred
Thinwall Polyallomer Tubes; Size(mm):11 x 34, Vol.(mL): 2.2
Catalog No. NC9780632 Supplier Beckman Coulter Supplier No. 347357
Thinwall Polyallomer Tubes; Size(mm):11 x 34, Vol.(mL): 2.2