Learn More
MSC Value Collection Poly Tubing, 12 in, 6 mil, 725 ft
Catalog No. 501917453
Encompass
Poly tubing provides the luxury of a custom fit poly bag and the flexibility of a wide variety of bag sizes without the expense. It is made from opaque, all virgin poly, which is suitable for light-sensitive products. The black, low-density polyethylene completely conceals contents.
  • Allows custom bags to be made by cutting tubing to desired length, inserting product, and heat sealing both ends.
  • Extra heavy-duty for puncture and tear resistance.
  • Low-density polyethylene completely conceals contents.
  • Opaque, all virgin poly is great for light-sensitive products.
Catalog No. 50-191-7453 Supplier MSC Supplier No. 39570239
Poly tubing provides the luxury of a custom fit poly bag and the flexibility of a wide variety of bag sizes without the expense. It is made from opaque, all virgin poly, which is suitable for light-sensitive products. The black, low-density polyethylene completely conceals contents.
  • Allows custom bags to be made by cutting tubing to desired length, inserting product, and heat sealing both ends.
  • Extra heavy-duty for puncture and tear resistance.
  • Low-density polyethylene completely conceals contents.
  • Opaque, all virgin poly is great for light-sensitive products.