Learn More
MSC Value Collection Poly Tubing, 14 in., 1.5 mil, 2900 ft
Catalog No. 501917391
Encompass
This clear poly tubing provides a versatile and cost-effective solution for creating custom-sized bags on demand. It offers the flexibility of a wide variety of bag sizes without the expense associated with pre-sized bags, suitable for numerous packaging applications.
  • Cut to desired length and heat seal both ends to create custom bags
  • Compatible with impulse sealers, bag tape, or twist ties for sealing
  • Meets FDA and USDA requirements for food application
  • Economical and lightweight, ideal for temporary storage of lightweight items
  • Dimensions are measured as width by length, representing inside measurements
Catalog No. 50-191-7391 Supplier MSC Supplier No. 39569561
This clear poly tubing provides a versatile and cost-effective solution for creating custom-sized bags on demand. It offers the flexibility of a wide variety of bag sizes without the expense associated with pre-sized bags, suitable for numerous packaging applications.
  • Cut to desired length and heat seal both ends to create custom bags
  • Compatible with impulse sealers, bag tape, or twist ties for sealing
  • Meets FDA and USDA requirements for food application
  • Economical and lightweight, ideal for temporary storage of lightweight items
  • Dimensions are measured as width by length, representing inside measurements