Learn More
MSC Value Collection Poly Tubing, 16 in, 6 mil, 725 ft
Catalog No. 501917454
Encompass
Poly Tubing provides the versatility of custom-fit poly bags and a wide range of sizes without the associated expense. Made from opaque, virgin low-density polyethylene, it effectively conceals contents and is suitable for light-sensitive products.
  • Custom bags can be made by cutting to desired length and heat sealing
  • Can be sealed using impulse sealers, bag tape, or twist ties
  • Extra heavy-duty 6 mil thickness for puncture and tear resistance
Catalog No. 50-191-7454 Supplier MSC Supplier No. 39570247
Poly Tubing provides the versatility of custom-fit poly bags and a wide range of sizes without the associated expense. Made from opaque, virgin low-density polyethylene, it effectively conceals contents and is suitable for light-sensitive products.
  • Custom bags can be made by cutting to desired length and heat sealing
  • Can be sealed using impulse sealers, bag tape, or twist ties
  • Extra heavy-duty 6 mil thickness for puncture and tear resistance