Learn More
MSC Value Collection Poly Tubing, 18 in, 3 mil, 1450 ft
Catalog No. 501917413
Encompass
Poly Tubing offers the luxury of a custom fit poly bag and the flexibility of a wide variety of bag sizes without the expense. It allows you to make custom bags in seconds by cutting to the desired length, inserting the product, and heat sealing both ends.
  • Custom fit poly bag
  • Flexibility of a wide variety of bag sizes
  • Make custom bags in seconds by cutting to desired length, inserting product, and heat sealing both ends
  • Can be sealed using Impulse Sealers, Bag Tape, or Twist Ties
  • Meets FDA and USDA requirements for food application
  • Sturdy mil protects medium to heavier products
Catalog No. 50-191-7413 Supplier MSC Supplier No. 39569819
Poly Tubing offers the luxury of a custom fit poly bag and the flexibility of a wide variety of bag sizes without the expense. It allows you to make custom bags in seconds by cutting to the desired length, inserting the product, and heat sealing both ends.
  • Custom fit poly bag
  • Flexibility of a wide variety of bag sizes
  • Make custom bags in seconds by cutting to desired length, inserting product, and heat sealing both ends
  • Can be sealed using Impulse Sealers, Bag Tape, or Twist Ties
  • Meets FDA and USDA requirements for food application
  • Sturdy mil protects medium to heavier products