Learn More
MSC Value Collection Poly Tubing, 24 in, 2.0 mil, 2150 in
Catalog No. 501917405
Encompass
Poly Tubing offers the luxury of a custom fit poly bag and the flexibility of a wide variety of bag sizes without the expense. Custom bags can be made in seconds by cutting the tubing to the desired length, inserting the product, and heat sealing both ends. It can be sealed using Impulse Sealers, Bag Tape, or Twist Ties. The clear poly tubing meets FDA and USDA requirements for food applications.
  • Offers luxury of custom fit poly bag
  • Provides flexibility of wide variety of bag sizes without expense
  • Custom bags can be made in seconds by cutting to desired length, inserting product, and heat sealing
  • Can be sealed using impulse sealers, bag tape, or twist ties
  • Clear poly tubing meets FDA and USDA requirements for food applications
Catalog No. 50-191-7405 Supplier MSC Supplier No. 39569710
Poly Tubing offers the luxury of a custom fit poly bag and the flexibility of a wide variety of bag sizes without the expense. Custom bags can be made in seconds by cutting the tubing to the desired length, inserting the product, and heat sealing both ends. It can be sealed using Impulse Sealers, Bag Tape, or Twist Ties. The clear poly tubing meets FDA and USDA requirements for food applications.
  • Offers luxury of custom fit poly bag
  • Provides flexibility of wide variety of bag sizes without expense
  • Custom bags can be made in seconds by cutting to desired length, inserting product, and heat sealing
  • Can be sealed using impulse sealers, bag tape, or twist ties
  • Clear poly tubing meets FDA and USDA requirements for food applications