Learn More
MSC Value Collection Poly Tubing, 2 in, 2 mil, 2150 ft
Catalog No. 501917399
Encompass
Poly Tubing allows for custom-fit poly bags and offers flexibility in bag sizes without additional expense. To create custom bags, simply cut the tubing to the desired length, insert the product, and heat seal both ends using Impulse Sealers, Bag Tape, or Twist Ties.
  • Creates custom-fit poly bags
  • Offers flexibility for various bag sizes
  • Easily cut to desired length
  • Heat sealable at both ends
  • Compatible with Impulse Sealers, Bag Tape, or Twist Ties
  • Clear tubing meets FDA and USDA requirements for food applications
  • Suitable for light to medium weight products (2 mil thickness)
Catalog No. 50-191-7399 Supplier MSC Supplier No. 39569652
Poly Tubing allows for custom-fit poly bags and offers flexibility in bag sizes without additional expense. To create custom bags, simply cut the tubing to the desired length, insert the product, and heat seal both ends using Impulse Sealers, Bag Tape, or Twist Ties.
  • Creates custom-fit poly bags
  • Offers flexibility for various bag sizes
  • Easily cut to desired length
  • Heat sealable at both ends
  • Compatible with Impulse Sealers, Bag Tape, or Twist Ties
  • Clear tubing meets FDA and USDA requirements for food applications
  • Suitable for light to medium weight products (2 mil thickness)