Learn More
MSC Value Collection Poly Tubing, 6 in, 4 mil, 1075 ft
Catalog No. 501920748
Encompass
Polyethylene tubing offers the flexibility to create custom-sized bags, reducing waste and cost. It can be cut to any desired length and sealed for immediate use.
  • Customizable length for poly bags
  • Can be sealed with impulse sealers, bag tape, or twist ties
  • Meets FDA and USDA requirements for food applications
  • Heavy-duty 4 mil thickness for industrial hardware and tools
Catalog No. 50-192-0748 Supplier MSC Supplier No. 39528922
Polyethylene tubing offers the flexibility to create custom-sized bags, reducing waste and cost. It can be cut to any desired length and sealed for immediate use.
  • Customizable length for poly bags
  • Can be sealed with impulse sealers, bag tape, or twist ties
  • Meets FDA and USDA requirements for food applications
  • Heavy-duty 4 mil thickness for industrial hardware and tools