Learn More
MSC Value Collection Poly Tubing, 6 in, 6 mil, 725 ft
Catalog No. 501917432
Encompass
This poly tubing provides a cost-effective solution for creating custom-sized bags suitable for various applications. It can be easily cut to the desired length, filled, and sealed to protect contents.
  • Easily create custom length bags
  • Compatible with impulse sealers, bag tape, and twist ties for sealing
  • Clear tubing meets FDA and USDA requirements for food applications
  • Extra heavy-duty 6 mil thickness for puncture and tear resistance
Catalog No. 50-191-7432 Supplier MSC Supplier No. 39570023
This poly tubing provides a cost-effective solution for creating custom-sized bags suitable for various applications. It can be easily cut to the desired length, filled, and sealed to protect contents.
  • Easily create custom length bags
  • Compatible with impulse sealers, bag tape, and twist ties for sealing
  • Clear tubing meets FDA and USDA requirements for food applications
  • Extra heavy-duty 6 mil thickness for puncture and tear resistance