Learn More
MSC Value Collection Poly Tubing, 8 in, 4 mil, 725 ft
Catalog No. 501917449
Encompass
Poly tubing allows for the creation of custom-fit poly bags, offering versatile sizing options without the expense of pre-sized bags.
  • Opaque, all virgin poly is suitable for light-sensitive products
  • Black, low-density polyethylene completely conceals contents
  • Custom bags can be made by cutting to desired length, inserting product, and heat sealing both ends
  • Can be sealed using impulse sealers, bag tape, or twist ties
  • Heavy-duty 4 mil thickness suitable for industrial hardware and robust protection
Catalog No. 50-191-7449 Supplier MSC Supplier No. 39570197
Poly tubing allows for the creation of custom-fit poly bags, offering versatile sizing options without the expense of pre-sized bags.
  • Opaque, all virgin poly is suitable for light-sensitive products
  • Black, low-density polyethylene completely conceals contents
  • Custom bags can be made by cutting to desired length, inserting product, and heat sealing both ends
  • Can be sealed using impulse sealers, bag tape, or twist ties
  • Heavy-duty 4 mil thickness suitable for industrial hardware and robust protection