Learn More
MSC Value Collection Poly Tubing, 8 in, 6 mil, 725 ft
Catalog No. 501920712
Encompass
Poly Tubing offers the luxury of a custom fit poly bag and the flexibility of a wide variety of bag sizes without the expense. Custom bags can be made in seconds by cutting the tubing to the desired length, inserting the product, and heat sealing both ends. This clear poly tubing meets FDA and USDA requirements for food applications, providing extra heavy-duty puncture and tear resistance.
  • Custom bag creation from continuous roll
  • Versatile, cut to any desired length and sealed
  • Compatible with impulse sealers, bag tape, or twist ties
  • Meets FDA and USDA requirements for food applications
  • Extra heavy-duty 6 mil thickness for puncture and tear resistance
Catalog No. 50-192-0712 Supplier MSC Supplier No. 39528799
Poly Tubing offers the luxury of a custom fit poly bag and the flexibility of a wide variety of bag sizes without the expense. Custom bags can be made in seconds by cutting the tubing to the desired length, inserting the product, and heat sealing both ends. This clear poly tubing meets FDA and USDA requirements for food applications, providing extra heavy-duty puncture and tear resistance.
  • Custom bag creation from continuous roll
  • Versatile, cut to any desired length and sealed
  • Compatible with impulse sealers, bag tape, or twist ties
  • Meets FDA and USDA requirements for food applications
  • Extra heavy-duty 6 mil thickness for puncture and tear resistance