Learn More
MSC Value Collection Poly Tubing, 9 in., 2 mil, 2150 ft.
Catalog No. 501917401
Encompass
Poly tubing offers the luxury of a custom fit poly bag and the flexibility of a wide variety of bag sizes without the expense. Custom bags can be created by cutting the tubing to the desired length, inserting the product, and heat sealing both ends.
  • Offers custom fit poly bags
  • Allows for flexible bag sizes
  • Can be sealed using impulse sealers, bag tape, or twist ties
  • Meets FDA and USDA requirements for food applications
Catalog No. 50-191-7401 Supplier MSC Supplier No. 39569678
Poly tubing offers the luxury of a custom fit poly bag and the flexibility of a wide variety of bag sizes without the expense. Custom bags can be created by cutting the tubing to the desired length, inserting the product, and heat sealing both ends.
  • Offers custom fit poly bags
  • Allows for flexible bag sizes
  • Can be sealed using impulse sealers, bag tape, or twist ties
  • Meets FDA and USDA requirements for food applications