Learn More
MSC Value Collection Poly Tubing, 9 in, 1.5 mil, 2900 ft
Catalog No. 501917392
Encompass
Poly tubing allows for the creation of custom-sized poly bags by cutting the tubing to the desired length, inserting the product, and heat sealing both ends, providing flexibility for various bag sizes without additional expense.
  • Can be sealed using impulse sealers, bag tape, or twist ties
  • Clear poly tubing meets FDA and USDA requirements for food applications
  • Thickness of 1.5 mil suitable for economical, lightweight, temporary storage of lightweight items
Catalog No. 50-191-7392 Supplier MSC Supplier No. 39569579
Poly tubing allows for the creation of custom-sized poly bags by cutting the tubing to the desired length, inserting the product, and heat sealing both ends, providing flexibility for various bag sizes without additional expense.
  • Can be sealed using impulse sealers, bag tape, or twist ties
  • Clear poly tubing meets FDA and USDA requirements for food applications
  • Thickness of 1.5 mil suitable for economical, lightweight, temporary storage of lightweight items