Learn More
MSC Miller Python Ultra Full-Body Harness, Vest-Style, 400 lb. Capacity, 1 ea.
Catalog No. 5000508647
Encompass
This full-body, vest-style harness is engineered for demanding applications including construction and confined space work. Constructed with stretchable polyester and urethane elastomer webbing, it is designed for a comfortable, secure fit while supporting a maximum working load of 400 lb (181.4 kg). It features quick-connect chest and leg strap buckles with a dual-tab release mechanism to facilitate easy, tangle-free donning.
  • Features a comfort-touch back D-ring pad for breathable, extended wear
  • Equipped with a sliding back D-ring and sub-pelvic strap for weight distribution
  • Includes tool belt loops and an integrated lanyard ring to minimize snag hazards
  • Includes a fall indicator that deploys to signal when the unit must be removed from service
  • Meets or exceeds OSHA and CSA specifications
  • Hardware meets ASTM salt spray test requirements
Catalog No. 50-005-08647 Supplier MSC Supplier No. 32279390
This full-body, vest-style harness is engineered for demanding applications including construction and confined space work. Constructed with stretchable polyester and urethane elastomer webbing, it is designed for a comfortable, secure fit while supporting a maximum working load of 400 lb (181.4 kg). It features quick-connect chest and leg strap buckles with a dual-tab release mechanism to facilitate easy, tangle-free donning.
  • Features a comfort-touch back D-ring pad for breathable, extended wear
  • Equipped with a sliding back D-ring and sub-pelvic strap for weight distribution
  • Includes tool belt loops and an integrated lanyard ring to minimize snag hazards
  • Includes a fall indicator that deploys to signal when the unit must be removed from service
  • Meets or exceeds OSHA and CSA specifications
  • Hardware meets ASTM salt spray test requirements