Learn More
MSC Miller DuraFlex Python Quick-Connect Harness, Dacron Polyester, 400 lb Capacity, Size S/M, 1 ea
Catalog No. 5000507195
Encompass
This stretchable, full-body harness is designed for use in industrial environments, offering dependable fall protection with a 400 lb. capacity. The harness features a sliding back D-ring, a sub-pelvic strap, and tool belt loops, constructed from high-strength materials to ensure both safety and durability.
  • Features DuraFlex® stretch webbing in the lower torso for improved comfort
  • Includes Quick-Connect chest and leg buckles for easy and efficient adjustments
  • Equipped with a Comfort-Touch back D-ring pad for better engagement
  • Designed with five points of adjustment for a secure, custom fit
  • Constructed with abrasion-resistant, 1-3/4 in. Dacron polyester webbing
  • Meets or exceeds all OSHA, ANSI, and CSA specifications
  • All hardware meets ASTM 50-hour salt spray test requirements
Catalog No. 50-005-07195 Supplier MSC Supplier No. 81379737
This stretchable, full-body harness is designed for use in industrial environments, offering dependable fall protection with a 400 lb. capacity. The harness features a sliding back D-ring, a sub-pelvic strap, and tool belt loops, constructed from high-strength materials to ensure both safety and durability.
  • Features DuraFlex® stretch webbing in the lower torso for improved comfort
  • Includes Quick-Connect chest and leg buckles for easy and efficient adjustments
  • Equipped with a Comfort-Touch back D-ring pad for better engagement
  • Designed with five points of adjustment for a secure, custom fit
  • Constructed with abrasion-resistant, 1-3/4 in. Dacron polyester webbing
  • Meets or exceeds all OSHA, ANSI, and CSA specifications
  • All hardware meets ASTM 50-hour salt spray test requirements