Learn More
Research Products International Corp Replacement Polypropylene Vial Caps, 22mm Poly Lined Plastic Screw Caps, White, 1000 per Package
SDP
Catalog No. 50213012
Encompass
Smooth top white caps available with various type liners for liquid scintillation counting applications. Recommended for all samples that have a pH of 5-7. For highly alkaline or acidic samples an inert liner is recommended.
Polypropylene Vial Caps, 22 mm Soft Poly Lined, White, Plastic Screw Cap
Catalog No. 50-213-012 Supplier Research Products International Corp Supplier No. 131003
Smooth top white caps available with various type liners for liquid scintillation counting applications. Recommended for all samples that have a pH of 5-7. For highly alkaline or acidic samples an inert liner is recommended.
Polypropylene Vial Caps, 22 mm Soft Poly Lined, White, Plastic Screw Cap