Learn More
Electron Microscopy Sciences Poly-View for Negatives, 35 mm
SDP
Catalog No. 5027965
Encompass_Preferred
Completely transparent polyethylene storage sheets fits all U.S. standard 3-ring binders: 8 1/4"x1 1/4". Convenient. Offers good protection for your valuable negatives and prints.
Size: 35 mm
Catalog No. 50-279-65 Supplier Electron Microscopy Sciences Supplier No. 74025
Completely transparent polyethylene storage sheets fits all U.S. standard 3-ring binders: 8 1/4"x1 1/4". Convenient. Offers good protection for your valuable negatives and prints.
Size: 35 mm