Learn More
Electron Microscopy Sciences Poly-View for Negatives, 4 x 5"
SDP
Supplier:  Electron Microscopy Sciences 74026
Encompass_Preferred
Completely transparent polyethylene storage sheets fits all U.S. standard 3-ring binders: 8 1/4"x1 1/4". Convenient. Offers good protection for your valuable negatives and prints.
Size: 4 x 5"
Catalog No. 50-279-63