Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(vinyl alcohol)

Catalog No. 3484061KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-840-61KG 1 kg
34-840-625G 25 g
34-840-6500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 34-840-61KG Supplier Sigma Aldrich Supplier No. 3484061KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(vinyl alcohol) (PVA) is a vinyl based water soluble polymer that can be used as a non-ionic surface active agent. It can also be used as a biodegradable polymer and can be incorporated in adhesives, coatings, textiles, ceramics and cosmetics. Its properties include good transparency, chemical resistance, toughness and antielectrostatic properties.PVA can be used for a variety of applications such as: a nanoparticle stabilizer for the surface modification of titanium surfacesin the preparation of an electrolyte for supercapacitorsformation of a composite with carbon nanotubes (CNTs) for thin film devices and flexible electronics

Specifications

CAS 9002-89-5
Linear Formula [-CH2CHOH-]n
MDL Number MFCD00081922
Quantity 1 kg
Molecular Weight (g/mol) 13,000-23,000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.