Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(vinyl alcohol-co-ethylene)

Catalog No. 414093100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-409-3100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-409-3100G Supplier Sigma Aldrich Supplier No. 414093100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(vinyl alcohol-co-ethylene) (PVA-co-PE) is a hydrophilic biocompatible polymer that has a non-toxic nature. It has high tensile strength and good film forming ability that make it useful in the food packaging industry.PVA-co-PE can be used as a membrane in a variety of applications like filtration, biosensor and food sensors. It can also be used as a nanoporous membrane, which can potentially be used as a separator in energy vehicles and lithium ion batteries.Maximum working temperature 240°C

Specifications

CAS 25067-34-9
Density 1.19 g/mL (at 25°C)
Linear Formula (CH2CH2)x[CH2CH(OH)]y
MDL Number MFCD00197931
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.