Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(vinyl Alcohol), Sigma-Aldrich

Catalog No. 01258907 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-907 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-907 Supplier MilliporeSigma Supelco Supplier No. P17631KG
Add to Cart
Add to Cart

Poly(vinyl alcohol) has been used in the preparation of multilayer active film for its antimicrobial analysis. It has also been used to study the effect of protein supplements on porcine parthenotes.

Poly(vinyl alcohol) is also suitable for:in vitro production (IVP) of bovine embryosoocyte recovery and in vitro maturationas a macromolecular component in basal cultured medium.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-89-5

Specifications

Quantity 1 kg
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.