Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(vinyl butyral-co-vinyl alcohol-co-vinyl acetate)

Catalog No. 182567250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-256-7250G 250 g
18-256-7500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-256-7250G Supplier Sigma Aldrich Supplier No. 182567250G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(vinyl butyral-co-vinyl alcohol-co-vinyl acetate)

Specifications

CAS 27360-07-2
Density 1.083 g/mL (at 25°C)
Refractive Index n20/D 1.485
MDL Number MFCD00132687
Quantity 250 g
Synonym Poly(vinyl butyral-vinyl alcohol-vinyl acetate)
Molecular Weight (g/mol) 50,000-80,000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.