Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(vinyl ethyleether), low molecular weight, Thermo Scientific™

Catalog No. AC179072500
Change view
Click to view available options
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC179072500 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC179072500 Supplier Thermo Scientific Chemicals Supplier No. 179072500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 25104-37-4
Molecular Formula C4H8O
Linear Formula {-CH2CH(OC2H5)-}n
MDL Number MFCD00009248
Density 96%g/mL
Synonym ethyl vinyl ether, ethoxyethene, ethoxyethylene, ethene, ethoxy, 1-ethoxyethene, 1-ethoxyethylene, vinyl ethyl ether, vinamar, ether, ethyl vinyl, ether, vinyl ethyl
Quantity 250g
InChI Key FJKIXWOMBXYWOQ-UHFFFAOYSA-N
SMILES CCOC=C
IUPAC Name ethoxyethene
Molecular Weight (g/mol) 72.11
PubChem CID 8023
Infrared Spectrum Authentic
Loss on Drying 10% max.
Chemical Name or Material Poly(vinyl ethyleether)
Show More Show Less
RTECSNumber CQ6693000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.