Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(vinyl Methyl Ether) (50% in Methanol), TCI America™
SDP

Catalog No. P1007500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1007500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1007500G Supplier TCI America Supplier No. P1007500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 9003-09-2
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00161528
InChI Key XJRBAMWJDBPFIM-UHFFFAOYSA-N
Synonym Methyl Vinyl Ether Polymer, Poly(methyl Vinyl Ether), Vinyl Methyl Ether Polymer
PubChem CID 7861
IUPAC Name methoxyethene
SMILES COC=C

Specifications

Quantity 500 g
UN Number 1230
Physical Form Liquid
Chemical Name or Material Poly(vinyl Methyl Ether) (50% in Methanol)
EINECSNumber (6)-0982
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.