Learn More
Research Products International Corp Pour-Boat Disposable Polystyrene Weighing Dishes, 3 1/2 x 5 1/4 x 1 Inches, 250 per Package
SDP
Catalog No. 50998463
Encompass
New design polystyrene weigh dish facilitates pouring and dispensing. Noneed to flex dish to dispense liquids or solids; 20-30% thickermaterial for strength, stability, handling ease and safety. New weigh dishes have a flat bottom and will withstand 200?F.They aredisposable, smooth, uniform and inexpensive.
Catalog No. 50-998-463 Supplier Research Products International Corp Supplier No. 146315
New design polystyrene weigh dish facilitates pouring and dispensing. Noneed to flex dish to dispense liquids or solids; 20-30% thickermaterial for strength, stability, handling ease and safety. New weigh dishes have a flat bottom and will withstand 200?F.They aredisposable, smooth, uniform and inexpensive.