Learn More
Kyfora Bio Poly(2-ethyl-2-oxazoline) [MW 500,000] 100 GR
Supplier:  Kyfora Bio 17810100
Encompass_Preferred
Poly(2-ethyl-2-oxazoline) is an amorphous water soluble polymer. It has been shown to have high biocompatibility, low accumulation in tissue, and rapid clearance from the bloodstream. It can also be used for formulation of extended release tablets for drug delivery. Key application(s): drug delivery
Catalog No. 50-255-9789