Learn More
Aldevron Poly(A) Polymerase, 3000ug
Catalog No. 503073887
Encompass_Preferred
3000ug dispense of Poly(A) Polymerase. Poly(A) Polymerase catalyzes addition of AMP from ATP to the 3' end of RNA in a template-independent manner.
Catalog No. 50-307-3887 Supplier Aldevron Supplier No. 91333000MCG
3000ug dispense of Poly(A) Polymerase. Poly(A) Polymerase catalyzes addition of AMP from ATP to the 3' end of RNA in a template-independent manner.