Learn More
Polysciences, Inc. Poly(acrylamide/acrylic acid)
SDP
Catalog No. NC1929718
Encompass
Anionic acrylamide polymer.90:10, Na saltViscosity:10% Aq 150-800 cps
Catalog No. NC1929718 Supplier Polysciences, Inc. Supplier No. 04652250
Anionic acrylamide polymer.90:10, Na saltViscosity:10% Aq 150-800 cps