Learn More
Lovibond™ Polyacrylate Reagent Set
Catalog No. 09050173 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
65 mL
Catalog No. Volume (Metric)
09-050-173 65 mL
1 options
Catalog No. 09-050-173 Supplier Lovibond™ Supplier No. 56R019165
Add to Cart
Add to Cart
Polyacrylate Reagent Set comes with 1x Polyacrylate Buffer A1 and Polyacrylate Precipitant A2 reagent for use with Lovibond method M338.
  • Commercially available C18 SPE cartridge and isopropyl alcohol are also required for the Polyacrylate test
Specifications
pH Range 0 to 14
Volume (Metric) 65 mL
Type Polyacrylate Reagent Set