Learn More
Polysciences, Inc. Poly(acrylic acid), Powder, Mw ~ 450,000 (PAA 450K) 100g
SDP
Catalog No. NC9278868
Encompass
Poly(acrylic acid), Composition: 2-Propenoic Acid, Homopolymer (9003-01-4); Acrylic Acid, MF: (C3H4O2)x, MW: ~1000000, n20/D: 1.527, FlashPoint: > 200 deg.F, SG: 1.41, pH : 2.5 to 3 1%H2O, WhitePowder, Store at RT, WaterSoluble, 100g
Catalog No. NC9278868 Supplier Polysciences, Inc. Supplier No. 03312100
Poly(acrylic acid), Composition: 2-Propenoic Acid, Homopolymer (9003-01-4); Acrylic Acid, MF: (C3H4O2)x, MW: ~1000000, n20/D: 1.527, FlashPoint: > 200 deg.F, SG: 1.41, pH : 2.5 to 3 1%H2O, WhitePowder, Store at RT, WaterSoluble, 100g