Learn More
Polysciences, Inc. Poly(acrylic acid), 63% soln. in water [MW ~2,000], 250g
SDP
Catalog No. NC9285082
Encompass_Preferred
Poly(acrylic acid), 63% soln. in water [MW ~2,000], Important anionic water-soluble polymer. Can be crosslinked covalently or ionically to form hydrogels. Mw/Mn 2.4, 63% soln. in water (157.5 g polymer), 9003-01-4, 250g
Catalog No. NC9285082 Supplier Polysciences, Inc. Supplier No. 06513250
Poly(acrylic acid), 63% soln. in water [MW ~2,000], Important anionic water-soluble polymer. Can be crosslinked covalently or ionically to form hydrogels. Mw/Mn 2.4, 63% soln. in water (157.5 g polymer), 9003-01-4, 250g