Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyacrylic Acid Sodium Salt, 45% Solution, Spectrum™ Chemical
SDP

Catalog No. 18605292 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-292 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-605-292 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1704500GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P1704, 9003-04-7, [CH2CH(CO2Na)]n

Spectrum™ Chemical Polyacrylic Acid Sodium Salt, 45 Percent Solution , also known as PAAc or Acrysol, is an anionic polymer used in disposable diapers and as thickening and emulsifying agents in the pharmaceutical and cosmetic industries. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9003-04-7 , 7732-18-5
Molecular Formula C3H3NaO2
Molecular Weight (g/mol) 94.05
InChI Key NNMHYFLPFNGQFZ-UHFFFAOYSA-M
IUPAC Name Sodium prop-2-enoate
SMILES [Na+].[O-]C(=O)C=C

Specifications

Color Viscous liquid
CAS Max % 0.45, 0.55
Quantity 500 g
Packaging Amber Glass Bottle
Viscosity 350-1000 cSt
Grade Ungraded
Chemical Name or Material Polyacrylic Acid Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.