Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Polyacrylic Acid Standards Kit, 10 GPC, 1,000 to 1,000,000 Daltons

Catalog No. 50785374 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This kit provides a collection of 10 Gel Permeation Chromatography (GPC) standards. It is specifically formulated for aqueous molecular weight determination and helps verify the performance of LC systems prior to GPC analysis.

  • 10 GPC standards
  • Molecular weight range of 1,000 to 1,000,000 Daltons
  • Supports gel permeation chromatography (GPC) for aqueous molecular weight determination and LC system performance verification
  • Solid formulation
  • Storage condition of 4°C (fridge)
  • Shelf life of 36 months; 1 week upon reconstitution
  • UV, RI/ELSD detection method
  • Compatible with LC system type
  • 10% tolerance (Mp)
  • 0.25 g mass
  • 10 units per package
  • GPC/GFC/SEC separation mode
  • Ambient shipping condition
  • Calibration type

Catalog No. 50-785-374 Supplier Waters Corp Supplier No. WAT035714
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This kit provides a collection of 10 Gel Permeation Chromatography (GPC) standards. It is specifically formulated for aqueous molecular weight determination and helps verify the performance of LC systems prior to GPC analysis.

  • 10 GPC standards
  • Molecular weight range of 1,000 to 1,000,000 Daltons
  • Supports gel permeation chromatography (GPC) for aqueous molecular weight determination and LC system performance verification
  • Solid formulation
  • Storage condition of 4°C (fridge)
  • Shelf life of 36 months; 1 week upon reconstitution
  • UV, RI/ELSD detection method
  • Compatible with LC system type
  • 10% tolerance (Mp)
  • 0.25 g mass
  • 10 units per package
  • GPC/GFC/SEC separation mode
  • Ambient shipping condition
  • Calibration type

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.