Learn More
Medchemexpress LLC Polyadenylic acid potassium | 26763-19-9 | MFCD00131937 | 90.0% | 385.31 g/mol | C10H13KN5O7P | 25 MG
SDP
Catalog No. 5000281222
Encompass_Preferred
Polyadenylic acid potassium (Poly(A)) is a polymeric adenylic acid potassium salt used to mimic the polyadenylate tail of eukaryotic mRNA. It is provided as a dry reagent for biochemical and biophysical assays, including mRNA stability, binding, and formulation studies.
  • Mimics the poly(A) tail for mRNA stability and binding assays.
  • Potassium salt form for compatibility with aqueous biochemical buffers.
  • Suitable for biochemical, biophysical, and formulation studies.
  • Available in research-scale quantities for laboratory use.
  • Manufacturer provides data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-81222 Supplier Medchemexpress LLC Supplier No. HYW41629125MG
Polyadenylic acid potassium (Poly(A)) is a polymeric adenylic acid potassium salt used to mimic the polyadenylate tail of eukaryotic mRNA. It is provided as a dry reagent for biochemical and biophysical assays, including mRNA stability, binding, and formulation studies.
  • Mimics the poly(A) tail for mRNA stability and binding assays.
  • Potassium salt form for compatibility with aqueous biochemical buffers.
  • Suitable for biochemical, biophysical, and formulation studies.
  • Available in research-scale quantities for laboratory use.
  • Manufacturer provides data sheet, certificate of analysis, and safety data sheet.