Learn More
Medchemexpress LLC Polyadenylic acid potassium | 26763-19-9 | MFCD00131937 | 90.0% | (C10H13KN5O7P)n | 50 MG
SDP
Catalog No. 5000281813
Encompass_Preferred
Polyadenylic acid potassium (Poly(A)) is the potassium salt of a homopolymeric adenylic acid that mimics the poly(A) tail of eukaryotic mRNA. It is used in molecular biology to study mRNA stability, protein-RNA interactions, and small-molecule binding to mRNA tails.
Catalog No. 50-002-81813 Supplier Medchemexpress LLC Supplier No. HYW41629150MG
Polyadenylic acid potassium (Poly(A)) is the potassium salt of a homopolymeric adenylic acid that mimics the poly(A) tail of eukaryotic mRNA. It is used in molecular biology to study mRNA stability, protein-RNA interactions, and small-molecule binding to mRNA tails.