Learn More
AdipoGen PolyADPrib PAR mAb 10H
SDP
Supplier:  AdipoGen AG20T0001C100
Encompass_Preferred
Monoclonal Antibody. Recognizes polyADP-ribose synthesized by a broad range of PARPs polyADP-ribose polymerases, including human, mouse, rat or Drosophila PARP enzymes. Isotype Mouse IgG3kappa. Clone 10H. Applications FACS, ICC, IHC, WB. Liquid. Containing 50mM HEPES, pH 7.4, 100mM NaCl, 1% BSA and 0.02% sodium azide. Processes such as transcription, repair and replication that require efficient DNA recognition are dependent on modulation of chromatin structure. Chromatin relaxation is a critical event that occurs during DNA repair and is associated with the negatively charged polymer of adenosine 5-diphosphate ADP-ribose PAR. PAR is synthesized from nicotinamide adenine dinucleotide NAD by the polyADP-ribose polymerase protein family PARPs, of which PARP-1 and to a lesser extent PARP-2 respond to DNA-strand breaks.
Catalog No. 50-256-3364