Learn More
Beckman Coulter POLYALLOMER TUBEX 11X34 T/W
Catalog No. NC9059640
Encompass_Preferred
Thickwall Polyallomer Tube, 11 x 34mm, Volume: 1.0mL, Qty.: 100
Catalog No. NC9059640 Supplier Beckman Coulter Supplier No. 347287
Thickwall Polyallomer Tube, 11 x 34mm, Volume: 1.0mL, Qty.: 100