Learn More
YMC America, Inc PolyamII 120A 5um 150x.3mm1/16
SDP
Catalog No. 5000878117
Encompass
YMC-Pack Polyamine II is a silica-based normal-phase stationary phase bonded with polyamine that is particularly useful for the separation of sugars The column lifetime in aqueous mobile phases is longer than conventional silica-based amino columns and is therefore applicable for the separation of oligosaccharides using high-aqueous mobile phases In addition YMC-Pack Polyamine II can be used to separate ionic compounds with a combination of normal-phase and weak anion-exchange modes
Catalog No. 50-008-78117 Supplier YMC America, Inc Supplier No. PB12S0515H0AU
YMC-Pack Polyamine II is a silica-based normal-phase stationary phase bonded with polyamine that is particularly useful for the separation of sugars The column lifetime in aqueous mobile phases is longer than conventional silica-based amino columns and is therefore applicable for the separation of oligosaccharides using high-aqueous mobile phases In addition YMC-Pack Polyamine II can be used to separate ionic compounds with a combination of normal-phase and weak anion-exchange modes