Learn More
YMC America, Inc PolyamII grd 120A 5um 10x4mm
SDP
Catalog No. 5000878714
Encompass
YMC-Pack Polyamine II is a silica-based normal-phase stationary phase bonded with polyamine that is particularly useful for the separation of sugars The column lifetime in aqueous mobile phases is longer than conventional silica-based amino columns and is therefore applicable for the separation of oligosaccharides using high-aqueous mobile phases In addition YMC-Pack Polyamine II can be used to separate ionic compounds with a combination of normal-phase and weak anion-exchange modes
Catalog No. 50-008-78714 Supplier YMC America, Inc Supplier No. PB12S050104GC
YMC-Pack Polyamine II is a silica-based normal-phase stationary phase bonded with polyamine that is particularly useful for the separation of sugars The column lifetime in aqueous mobile phases is longer than conventional silica-based amino columns and is therefore applicable for the separation of oligosaccharides using high-aqueous mobile phases In addition YMC-Pack Polyamine II can be used to separate ionic compounds with a combination of normal-phase and weak anion-exchange modes