Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Polyaniline | 25233-30-1 | MFCD00241446 | 98.6% | C8H11N | 100 G
SDP

Supplier:  Medchemexpress LLC HYW017767100G

Encompass_Preferred

Polyaniline is a research-grade conductive polymer (CAS 25233-30-1) used in materials science and applied chemistry research, including development of antibacterial agents, sensors, coatings, and conductive composites. Reported properties include a molecular formula of C8H11N and a supplier-listed purity of 98.6%. Documentation such as a data sheet, certificate of analysis, and safety data sheet are available.

  • Conductive polymer for materials and electronics research.
  • Reported purity of 98.6% suitable for research applications.
  • Molecular formula C8H11N reflects the monomeric repeat unit.
  • Applicable as a precursor or component in antibacterial agent development.
  • Available in common laboratory package sizes, including 100 g.
  • Documentation available: data sheet, COA, and SDS.

Catalog No. 50-003-01644


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.