Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polyaniline (emeraldine base)

Catalog No. 5763795G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-637-95G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-637-95G Supplier Sigma Aldrich Supplier No. 5763795G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Polyaniline (emeraldine base) (PANI-EB) is an intrinsically conducting polymer (ICP) that can be formed by oxidative polymerization of aniline. Emeraldine base contains oxidized diimine and reduced diamine. PANI can be modified into the conjugating emeraldine base form by p-type doping with protonic acid.PANI-EB can form a nanocomposite with multi-walled carbon nanotubes (MWCNTs) that can be used to functionalize glassy carbon electrodes for the electro-sensitive detection of cadmium (Cd(II)) and lead (Pb(II)). Carbon nanofibers may be reinforced on PANI-EB to yield a specific capacitance of 95 F/g and a 92% retention over 3000 cycles as observed in supercapacitor applications. It may be used with heptamethyleneimine as an inhibitor to form stable fuel cell membranes.

Specifications

CAS 25233-30-1
MDL Number MFCD00241446
Quantity 5 g
Synonym Emeraldine base polyaniline
Molecular Weight (g/mol) 100000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.