Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polyaniline (emeraldine base)

Catalog No. 5563785G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-637-85G 5 g
55-637-825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 55-637-85G Supplier Sigma Aldrich Supplier No. 5563785G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Polyaniline (emeraldine base) (PANI-EB) is a conductive polymer that can be chemically synthesized from aniline by oxidative polymerization of ammonium peroxydisulfate in acidic solution. It is widely used in electronics and optoelectronic applications.PANI-EB is a conjugating polymer that finds applications in organic light emitting diodes (OLEDs), anticorrosive coatings, chemical sensors and solar cells.

Specifications

CAS 25233-30-1
MDL Number MFCD00241446
Quantity 5 g
Synonym Emeraldine base polyaniline
Molecular Weight (g/mol) 20000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.