Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polyaniline (emeraldine base)

Catalog No. 5564595G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-645-95G 5 g
55-645-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 55-645-95G Supplier Sigma Aldrich Supplier No. 5564595G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Polyaniline (emeraldine base) (PANI) is anintrinsic conductive polymer which consists of a ring structure attached with nitrogen substituted derivatives. It has an emeraldine type oxidation state that exists in the form of its base. It can be prepared by deprotonation of emeraldine hydrochloric acid and ammonium hydroxide. It is mainly used in the development of electrochemical devices.PANI forms a conjugating system that can be used in semiconducting applications. It can be used in a variety of other applications which include protective coating, hole transporting layer for organic electronics and chemical sensing.

Specifications

CAS 25233-30-1
MDL Number MFCD00241446
Quantity 5 g
Synonym Emeraldine base polyaniline
Molecular Weight (g/mol) 5000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.