Learn More
HIMEDIA PolyB Selective Supplement, FD003, Microbiology Culture Media, Selective Supplement, Antibiotic Supplement, Selective Isolation, Lyophilized, Vials, 5 vials
Catalog No. NC4013612
Encompass_Preferred
PolyB Selective Supplement is a lyophilized antibiotic supplement that adds polymyxin B selectivity to base media for the recovery of target organisms such as Bacillus cereus. Each vial, sufficient for 500 to 1000 ml of medium, supplies polymyxin B sulphate, which suppresses competing Gram-negative flora so the organisms of interest stand out. Rehydrate with sterile distilled water and add to molten Bacillus Cereus Agar Base or a comparable medium. Store at 2 to 8 C. Packaged as 5 vials.
Catalog No. NC4013612 Supplier HIMEDIA Supplier No. FD0035VL
PolyB Selective Supplement is a lyophilized antibiotic supplement that adds polymyxin B selectivity to base media for the recovery of target organisms such as Bacillus cereus. Each vial, sufficient for 500 to 1000 ml of medium, supplies polymyxin B sulphate, which suppresses competing Gram-negative flora so the organisms of interest stand out. Rehydrate with sterile distilled water and add to molten Bacillus Cereus Agar Base or a comparable medium. Store at 2 to 8 C. Packaged as 5 vials.