Learn More
MSC Value Collection Polybag, 12 x 12 in, 6 mil, 500/Pk.
Catalog No. 501920652
Encompass
These reclosable polybags are designed to protect contents from moisture and maintain cleanliness, suitable for various storage and packaging needs. They are manufactured from 100% virgin, high clarity polyethylene film and meet FDA and USDA specifications.
  • Zipper closure extends the width of the bag
  • Can be reused
  • Six mil thickness for extra heavy-duty puncture and tear resistance
  • Clear film for easy content identification
  • Inside usable dimensions are 12 x 12 in
Catalog No. 50-192-0652 Supplier MSC Supplier No. 39532544
These reclosable polybags are designed to protect contents from moisture and maintain cleanliness, suitable for various storage and packaging needs. They are manufactured from 100% virgin, high clarity polyethylene film and meet FDA and USDA specifications.
  • Zipper closure extends the width of the bag
  • Can be reused
  • Six mil thickness for extra heavy-duty puncture and tear resistance
  • Clear film for easy content identification
  • Inside usable dimensions are 12 x 12 in