Learn More
MSC Value Collection Polybags, 4 mil, 28 x 36 in, 100/Pk
Catalog No. 501920740
Encompass
Poly bags provide a versatile and flexible storage solution for various applications, including food, industrial tools, and electronic equipment, effectively protecting contents from dirt, dust, and moisture.
  • Manufactured from 100% virgin, high clarity polyethylene film
  • Meets FDA and USDA specifications
  • Recyclable
  • Open top for closure with twist ties, bag tape, or an impulse sealer
Catalog No. 50-192-0740 Supplier MSC Supplier No. 39539721
Poly bags provide a versatile and flexible storage solution for various applications, including food, industrial tools, and electronic equipment, effectively protecting contents from dirt, dust, and moisture.
  • Manufactured from 100% virgin, high clarity polyethylene film
  • Meets FDA and USDA specifications
  • Recyclable
  • Open top for closure with twist ties, bag tape, or an impulse sealer