Learn More
MSC Value Collection Gusseted Bag, 38 in, 3 mil, 50 Pack
Catalog No. 501916988
Encompass
Value Collection gusseted bags are designed for protecting items from dirt, dust, and moisture during storage or transport. Featuring expandable sides and a fitted bottom, these bags are ideal for packing bulky or irregularly shaped products. With a 3 mil thickness, they provide sturdy protection for medium to heavier goods.
  • Expandable sides and fitted bottom for easy packing of bulky or irregular shaped items
  • Protects contents from dirt, dust, and moisture
  • Manufactured from 100% virgin, high clarity polyethylene film
  • Meets FDA and USDA specifications
  • Can be closed with twist ties, bag tape, or impulse sealer
  • Bags are recyclable
  • Sturdy 3 mil thickness for medium to heavier products
Catalog No. 50-191-6988 Supplier MSC Supplier No. 39565353
Value Collection gusseted bags are designed for protecting items from dirt, dust, and moisture during storage or transport. Featuring expandable sides and a fitted bottom, these bags are ideal for packing bulky or irregularly shaped products. With a 3 mil thickness, they provide sturdy protection for medium to heavier goods.
  • Expandable sides and fitted bottom for easy packing of bulky or irregular shaped items
  • Protects contents from dirt, dust, and moisture
  • Manufactured from 100% virgin, high clarity polyethylene film
  • Meets FDA and USDA specifications
  • Can be closed with twist ties, bag tape, or impulse sealer
  • Bags are recyclable
  • Sturdy 3 mil thickness for medium to heavier products