Learn More
MSC Value Collection Polybags, 9x12in, 2 mil, Black, 1000/Pk.
Catalog No. 501917284
Encompass
These reclosable poly bags feature a zipper that extends the width of the bag, effectively keeping products clean and free from moisture. Manufactured from 100% virgin, high clarity polyethylene film, they are reusable and meet both FDA and USDA specifications, making them suitable for a variety of applications.
  • Zipper extends width to keep products clean and free from moisture
  • Reusable design for repeated use
  • Manufactured from 100% virgin, high clarity polyethylene film
  • Meets FDA and USDA specifications
  • Dimensions listed represent the inside usable measurements of the bag
  • Features 2 mil thickness suitable for light to medium weight products
Catalog No. 50-191-7284 Supplier MSC Supplier No. 39568449
These reclosable poly bags feature a zipper that extends the width of the bag, effectively keeping products clean and free from moisture. Manufactured from 100% virgin, high clarity polyethylene film, they are reusable and meet both FDA and USDA specifications, making them suitable for a variety of applications.
  • Zipper extends width to keep products clean and free from moisture
  • Reusable design for repeated use
  • Manufactured from 100% virgin, high clarity polyethylene film
  • Meets FDA and USDA specifications
  • Dimensions listed represent the inside usable measurements of the bag
  • Features 2 mil thickness suitable for light to medium weight products