Learn More
Polysciences, Inc. Polybead Amino Microspheres 3.00 um, 5 mL
SDP
Catalog No. NC0585754
Encompass
Polybead Amino Microspheres are monodisperse polystyrene microspheres that contain surface primary amine groups. Protein coupling using glutaraldehyde as a coupling agent will result in protein binding 11-12 carbon atoms from the surface of the bead. These microspheres are packaged as 2.5% solids (w/v) aqueous suspensions.
Catalog No. NC0585754 Supplier Polysciences, Inc. Supplier No. 171455
Polybead Amino Microspheres are monodisperse polystyrene microspheres that contain surface primary amine groups. Protein coupling using glutaraldehyde as a coupling agent will result in protein binding 11-12 carbon atoms from the surface of the bead. These microspheres are packaged as 2.5% solids (w/v) aqueous suspensions.