Learn More
Polysciences, Inc. Polybead® Amino Sampler Kit
SDP
Catalog No. NC3876580
Encompass
Kit contains 5ml of amino polystyrene monodisperse microspheres (2.5% solids in water) for each of these nominal sizes: 0.5µm, 0.75µm, 1.0µm and 3.0µm.
Catalog No. NC3876580 Supplier Polysciences, Inc. Supplier No. 198201
Kit contains 5ml of amino polystyrene monodisperse microspheres (2.5% solids in water) for each of these nominal sizes: 0.5µm, 0.75µm, 1.0µm and 3.0µm.